EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Solid state thermal degradation products of rosaramicin.
PMID 1429241
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2636
Rosamicin, from Micromonospora rosaria, ~98%
C31H51NO9