EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
LIVER MICROSOMAL GLUCOSE 6-PHOSPHATASE, INORGANIC PYROPHOSPHATASE, AND PYROPHOSPHATE-GLUCOSE PHOSPHOTRANSFERASE. 3. ASSOCIATED NUCLEOSIDE TRIPHOSPHATE- AND NUCLEOSIDE DIPHOSPHATE-GLUCOSE PHOSPHOTRANSFERASE ACTIVITIES.
PMID 14299641