EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Determination of sterols, erythrodiol, uvaol and alkanols in olive oils using combined solid-phase extraction, high-performance liquid chromatographic and high-resolution gas chromatographic techniques.
PMID 1430013
Abstract
A method is described for the determination of the sterol, erythrodiol, uvaol and alkanol content in olive oils by means of solid-phase extraction and high-performance liquid chromatography, instead of liquid-liquid and thin-layer chromatographic separations, the following step being high-resolution gas chromatographic separation. This type of procedure allows the simultaneous analysis of a larger number of samples and a substantial reduction in manual operations. Comparisons were made between the two methods on 100 different olive oils and with a statistical analysis of the results (Student's t-test).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09258
Erythrodiol, analytical standard
C30H50O2
U6628
Uvaol, ≥95%
C30H50O2