EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Solid-phase extraction and HPLC determination of 4-vinyl guaiacol and its precursor, ferulic acid, in orange juice.
PMID 1430047
Abstract
This study is undertaken to develop a simplified, rapid method to determine both immediate and potential off odors due to 4-vinyl guaiacol and its odorless precursor, ferulic acid, from a single sample preparation and chromatographic analysis. Orange juice sample preparation consists of a simple, C18 solid phase extraction. Utilizing a 5-microns, 25-cm, C18 column, both compounds can be separated within 40 min using a one-step, linear gradient beginning with an aqueous 12% tetrahydrofuran (THF)-5% acetonitrile mixture and ending with 35% aqueous THF. Hesperidin and nariutin have been identified as the compounds that interfered with the ultraviolet (UV) determination of sinapic and caffeic acids. Fluorescence detection with wavelength programming offers optimal sensitivity and selectivity. Recoveries of 4-vinyl guaiacol and ferulic acid range from 90 to 103%. Detection limits are 1 ppm and 5 ppm for ferulic acid and 4-vinyl guaiacol, respectively. Other hydroxycinnamic acids such as coumaric, sinapic, and caffeic acids may also be determined from the same chromatogram.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W267511
2-Methoxy-4-vinylphenol, ≥98%, FG
C9H10O2