EMAIL THIS PAGE TO A FRIEND
Pharmazie in unserer Zeit
[The color of alkyl-diammonium-rhodizonate].
PMID 1438472
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
392340
Potassium rhodizonate, technical grade, 85%
C6K2O6