EMAIL THIS PAGE TO A FRIEND
Toxicon : official journal of the International Society on Toxinology
Antivenom and biological effects of ar-turmerone isolated from Curcuma longa (Zingiberaceae)
PMID 1440627
Abstract
A potent antivenom against snakebite was isolated from Curcuma longa, a plant commonly used in traditional Brazilian medicine. The fraction consisting of ar-turmerone neutralized both the hemorrhagic activity present in Bothrops jararaca venom, and the lethal effect of Crotalus durissus terrificus venom in mice. Immunological studies demonstrated that this fraction also inhibited the proliferation and the natural killer activity of human lymphocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42258
(S)-ar-Turmerone, analytical standard
C15H20O