EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Isocaproic acid, a metabolite of 20 alpha-hydroxycholesterol.
PMID 14446007
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W346306
4-Methylpentanoic acid, ≥98%, FCC, FG
C6H12O2
277827
4-Methylvaleric acid, 99%
C6H12O2