EMAIL THIS PAGE TO A FRIEND
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons
Ketorolac tromethamine: an oral/injectable nonsteroidal anti-inflammatory for postoperative pain control.
PMID 1447612
Abstract
Ketorolac tromethamine is a new injectable/oral nonsteroidal anti-inflammatory analgesic with no apparent opiate receptor activity that has been administered alone and in combination with other opiate analgesics for the treatment of postoperative pain. The drug has shown promise in analgesic comparisons with morphine sulfate; it lacks the effects of respiratory depression and nausea and vomiting usually associated with narcotic agents. Intramuscular ketorolac may be particularly useful with those patients who have respiratory disease and patients being dismissed following short ambulatory or private-office anesthetic procedures.