EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Enzymatic synthesis of polynucleotides. IV. Purification and properties of polynucleotide phosphorylase of Azotobacter vinelandii.
PMID 14481058