EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Alginic acid metabolism in bacteria. I. Enzymatic formation of unsaturated oligosac-charides and 4-deoxy-L-erythro-5-hexoseulose uronic acid.
PMID 14488584