EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
A zinc clasp structure tethers Lck to T cell coreceptors CD4 and CD8.
PMID 14500983
Abstract
The T cell coreceptors CD4 and CD8 both associate via their cytoplasmic tails with the N-terminus of the Src-family tyrosine kinase Lck. These interactions require zinc and are critical for T cell development and activation. We examined the folding and solution structures of ternary CD4-Lck-Zn2+ and CD8alpha-Lck-Zn2+ complexes. The coreceptor tails and the Lck N-terminus are unstructured in isolation but assemble in the presence of zinc to form compactly folded heterodimeric domains. The cofolded complexes have similar "zinc clasp" cores that are augmented by distinct structural elements. A dileucine motif required for clathrin-mediated endocytosis of CD4 is masked by Lck.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2752
Leu-Leu
C12H24N2O3