EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Superhydrophobicity of nanostructured carbon films in a wide range of pH values.
PMID 14502740
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
222380
Allylpalladium(II) chloride dimer, 98%
C6H10Cl2Pd2