EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Amplification of dynamic chiral crown ether complexes during cyclic acetal formation.
PMID 14502741
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
377619
D-Threitol, 99%
C4H10O4
263559
DL-Threitol, 97%
C4H10O4