EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and AMPA receptor antagonistic activity of a novel class of quinoxalinecarboxylic acid with a substituted phenyl group at the C-7 position.
PMID 14505662
Abstract
The synthesis and biological properties of a novel class of 7-heterocycle-substituted quinoxalinecarboxylic acids, which bear a substituted phenyl group through a urethane linkage at the C-7 position, are described. One of the synthesized compounds, 15l, which has a 4-carboxyphenyl carbamoyloxymethyl imidazole group at the C-7 position and is water-soluble, was found to possess high potency in vitro and showed excellent neuroprotective efficacy in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
293407
2-Quinoxalinecarboxylic acid, 97%
C9H6N2O2