EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Fungicide chinomethionate as a new family of photoinducible DNA-cleaving agents.
PMID 14505671
Abstract
It is demonstrated for the first time in this report that chinomethionate is capable of causing efficient DNA cleavage under mild irradiation conditions, a fungicide molecule that processes the simple group of 1,3-dithio-2-one as its reactive functionality.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45372
Chinomethionate, PESTANAL®, analytical standard
C10H6N2OS2