EMAIL THIS PAGE TO A FRIEND
Farmaco (Societa chimica italiana : 1989)
Chemo-enzymatic synthesis of levodropropizine.
PMID 14505734
Abstract
Levodropropizine, an antitussive drug, was prepared in high enantiomeric excess in three steps, starting from dichloroacetone (2). Monosubstitution of 2 with sodium benzoate and subsequent baker's yeast reduction stereoselectively afforded the corresponding chlorohydrin in 73% ee, which was converted to levodropropizine and enantiomerically enriched up to 95% ee by fractional crystallisation.