EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Effects of different dilution water types on the acute toxicity to juvenile Pacific salmon and rainbow trout of fluroxypyr formulated product XRM-5084.
PMID 1450574
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36780
Fluroxypyr-1-methylheptyl ester, PESTANAL®, analytical standard
C15H21Cl2FN2O3