EMAIL THIS PAGE TO A FRIEND
Melanoma research
Fotemustine: an overview of its clinical activity in disseminated malignant melanoma.
PMID 1450667
Abstract
Fotemustine is a new chloronitrosourea which is active against disseminated malignant melanoma (DMM), and especially against cerebral metastases (CM). This efficacy has been widely demonstrated through many phase II studies. A multicentre trial of monotherapy was undertaken in 153 evaluable French patients. A response rate (RR) of 24.2% (25.0% RR in CM; 31.8% in non-visceral metastases (NVM) was obtained. Three other phase II studies confirmed these results with respective objective RR of 16.7%, 20.0% and 47.0% and RR in CM of 8.3%, 14.3% and 60.0%. Fotemustine has also been used in combination with dacarbazine (DTIC), in patients with DMM. The RR among 103 patients was 27.2% (26.3% in CM, 37.5% in NVM), confirming the activity of fotemustine. The two drugs have also been administered sequentially, in order to exploit their synergism in interfering with the O6 alkyltransferase. Impressive RR have been achieved, especially in patients with visceral metastases (VM) but at the expense of a pulmonary toxicity that does not arise with other treatment schedules and which precludes its use outside of strictly conducted clinical trials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7307
Fotemustine, ≥98% (HPLC)
C9H19ClN3O5P