EMAIL THIS PAGE TO A FRIEND
Organic letters
Highly enantioselective catalytic phenylation of ketones with a constrained geometry titanium catalyst.
PMID 14507193
Abstract
[reaction: see text] The catalytic asymmetric addition of phenyl groups from diphenylzinc to ketones is reported. The catalyst, generated from a dihydroxy bis(sulfonamide) ligand and titanium tetraisopropoxide, gives good to excellent enantioselectivities with a range of substrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
233161
3′-(Trifluoromethyl)acetophenone, 99%
C9H7F3O
248193
3′-Chloropropiophenone, 98%
C9H9ClO