EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Determination of clopidol residues in chicken tissues by liquid chromatography: collaborative study.
PMID 14509425
Abstract
Eighteen laboratories participated in a collaborative study on the determination of clopidol residues in chicken muscle tissues by liquid chromatography. Of these, results from 16 laboratories which rigorously followed the method were subjected to statistical analysis. The method performance was assessed by all participants using 14 samples of chicken muscle fortified at concentrations ranging from 0.1 to 5.0 mg/kg. In addition, 9 participants each reported results for 6 clopidol-incurred samples in chicken muscle. Test portions were extracted with acetonitrile, and the extracts were purified with alumina and anion exchange resin solid-phase extraction cartridges in sequence. Clopidol was separated by reversed-phase liquid chromatography and quantified at 270 nm. Average recoveries ranged from 81.8 to 85.4%, reproducibility relative standard deviation (RSDR) ranged from 11.9 to 22.6%, and repeatability relative standard deviation (RSDr) ranged from 9.9 to 15.1%. For clopidol-incurred samples at concentrations of 0.100-0.687 mg/kg, the mean determination value range was 0.099-0.659 mg/kg; RSDR was 12.6-19.8%, RSDr was 3.1-8.5%; and HORRAT values were 0.7-1.1. The accuracy and precision of the method are in conformity with the requirements specified by AOAC INTERNATIONAL. The method was adopted Official First Action in April 2003.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33988
Clopidol, VETRANAL®, analytical standard
C7H7Cl2NO