EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Molecular structure and stereochemistry of silybin A, silybin B, isosilybin A, and isosilybin B, Isolated from Silybum marianum (milk thistle).
PMID 14510591
Abstract
Two pairs of diastereoisomeric flavonolignans, silybin A, silybin B, isosilybin A, and isosilybin B, were successfully separated from Silybum marianum by sequential silica gel column chromatography, preparative reversed-phase HPLC, and recrystallization. Complete stereochemical assignments at C-2, C-3, C-7', and C-8' of these flavonolignans have been achieved. On the basis of X-ray crystallographic analysis and optical rotation data, coupled with comprehensive (1)H and (13)C NMR spectral data interpretation including COSY, HMQC, and HMBC, the stereochemistry of these diastereoisomers was determined unambiguously as silybin A (4), 2R, 3R, 7'R, 8'R; silybin B (5), 2R, 3R, 7'S, 8'S; isosilybin A (6), 2R, 3R, 7'R, 8'R; and isosilybin B (7), 2R, 3R, 7'S, 8'S.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2383
Isosilybin, ≥98% (HPLC)
C25H22O10