EMAIL THIS PAGE TO A FRIEND
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete
[Anal and palmar contact dermatitis caused by iodopropynyl butylcarbamate in moist sanitary wipes].
PMID 14513245
Abstract
A 63-year old man developed severe perianal and palmar contact dermatitis caused by sensitization to iodopropynyl butylcarbamate in moist sanitary wipes. Iodopropynyl butylcarbamate is increasingly employed as preservative in common cosmetic formulations and moist sanitary wipes as substitute for the previously frequently used sensitizers isothiazolinones and methyldibromoglutaronitrile. The allergic potential of diverse preservatives is a serious problem for the cosmetic industry, since truly hypoallergenic preservatives are not available but products such as moist sanitary wipes in large packages definitely require some form of protection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
521949
3-Iodo-2-propynyl N-butylcarbamate, 97%
C8H12INO2