EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Occupational allergic contact dermatitis from ethyl chloro oximido acetate.
PMID 1451513
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
292621
Ethyl 2-chloro-2-(hydroxyimino)acetate, 97%
C4H6ClNO3