EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
A defect in the p53 response pathway induced by de novo purine synthesis inhibition.
PMID 14517211
Abstract
p53 is believed to sense cellular ribonucleotide depletion in the absence of DNA strand breaks and to respond by imposition of a p21-dependent G1 cell cycle arrest. We now report that the p53-dependent G1 checkpoint is blocked in human carcinoma cell lines after inhibition of de novo purine synthesis by folate analogs inhibitory to glycinamide ribonucleotide formyltransferase (GART). p53 accumulated in HCT116, MCF7, or A549 carcinoma cells upon GART inhibition, but, surprisingly, transcription of several p53 targets, including p21cip1/waf1, was impaired. The mechanism of this defect was examined. The p53 accumulating in these cells was nuclear but was not phosphorylated at serines 6, 15, and 20, nor was it acetylated at lysines 373 or 382. The DDATHF-stabilized p53 bound to the p21 promoter in vitro and in vivo but did not activate histone acetylation over the p53 binding sites in the p21 promoter that is an integral part of the transcriptional response mediated by the DNA damage pathway. We concluded that the robust initial response of the p53 pathway to GART inhibitors is not transcriptionally propagated to target genes due to a defect in p53 post-translational modifications and a failure to open chromatin structure despite promoter binding of this unmodified p53.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0040
Lometrexol hydrate, ≥95% (HPLC)
C21H25N5O6 · xH2O