EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Subsite structure of maltogenic amylase from Thermomonospora viridis TF-35.
PMID 14519988
Abstract
We estimated the subsite structure of the maltogenic amylase from Thermomonospora viridis TF-35 (TVA). TVA has six subsites, and the catalytic site is between the 3rd and 4th subsite. The subsite affinities, A(-3), A(-2), (A(-1)+A(+1)), A(+2) and A(+3) were calculated to be 0.47, 2.31, 0.49, 2.45, and 0.17 kcal mol(-1), respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2986
Amylase, Maltogenic from Bacillus sp.