EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of chlorophenols in soil samples by microwave-assisted extraction coupled to headspace solid-phase microextraction and gas chromatography-electron-capture detection.
PMID 14521307
Abstract
Microwave-assisted extraction coupled to headspace solid-phase microextraction was studied and applied for one-step in-situ sample preparation prior to analysis of chlorophenols (CPs) in soil samples. The CPs in soil sample were extracted into the aqueous solution and then directly onto the solid-phase microextraction (SPME) fiber in headspace under the aid of microwave irradiation. After being desorbed from SPME fiber in the GC injection port, CPs were analyzed with a GC-electron-capture detection system. Parameters affecting the extraction efficiency such as the extraction solutions, the pH in the slurry, the humic acid content in the soil, the power and the irradiation time of microwave as well as the desorption parameters were investigated. Experimental results indicated that the extraction of a 1.0 g soil sample with a 6-ml aqueous solution (pH 2) and a polyacrylate fiber under the medium-power irradiation (132 W) for 9 min achieved the best extraction efficiency of about 90% recovery and less than 10% RSD. Desorption was optimal at 300 degrees C for 3 min. Detection limits were obtained at around 0.1-2.0 microg/kg levels. The proposed method provided a simple, fast, and organic solvent-free procedure to analyze CPs from soil sample matrix.