EMAIL THIS PAGE TO A FRIEND
Natural product research
Stereospecific hydroxylation of (+)-cedrol by using human skin microbial flora Staphylococcus epidermidis.
PMID 14526908
Abstract
Microbial transformation of (+)-cedrol was investigated by using Staphylococcus epidermidis and found that stereospecific hydroxylation of (+)-cedrol occurred at the C-3 position to form (+)-(3S)-3-hydroxycedrol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
22135
(+)-Cedrol, ≥99.0% (sum of enantiomers, GC)
C15H26O
W521418
Cedrol, redistilled
C15H26O