EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Self-promoted electron transfer from cobalt(II) porphyrin to p-fluoranil to produce a dimer radical anion-cobalt(III) porphyrin complex.
PMID 14531678
Abstract
Self-promoted electron transfer from a cobalt(II) porphyrin [Co(II)OEP] to p-fluoranil (F4Q) occurs, exhibiting a second-order dependence of the electron-transfer rate with respect to the F4Q concentration due to the formation of a strong complex between the dimer radical anion [(F4Q)2*-] and the resulting Co(III)OEP+.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
104353
Tetrafluoro-1,4-benzoquinone, 97%
C6F4O2