EMAIL THIS PAGE TO A FRIEND
Physiological research
Frequency-domain lifetime fluorometry of double-labeled creatine kinase.
PMID 14535833
Abstract
Myofibril-bound creatine kinase EC 2.7.3.2 (CK), a key enzyme of muscle energy metabolism, has been selected for studies of conformational changes that underlie the cellular control of enzyme activity. For fluorescence spectroscopy measurements, the CK molecule was double-labeled with IAF (5-iodoacetamidofluorescein) and ErITC (erythrosin 5'-isothiocyanate). Measurement of fluorescence resonance energy transfer (FRET) from fluorescein to erythrosin was used to obtain information about the donor-acceptor pair distance. Frequency-domain lifetime measurements evaluate the donor-acceptor distance in the native CK molecule as 7.8 nm. The Förster radius equals 5.3 nm with the resolution range from 0.2 to 1.0 nm. Erythrosin-fluorescein labeling (EFL) was tested for artificial conformational changes of the CK molecule with high-salt concentration treatment. The transition distance, defined by His-97 and Cys-283 and derived from a 3D model equals 0.766 nm for the open (inactive) form and 0.277 nm for the closed (reactive) form of the CK molecule. In this way, the resolution range of the used spectroscopy method is significant, concerning the difference of 0.489 nm. Nevertheless, the CK enzyme activity, assessed by the hexokinase-coupled assay, was diminished down to 1 % of the activity of the native enzyme. EFL is suitable for description of conformational behavior implied from the regulation of creatine kinase. However, the observed inhibition restricts EFL to studies of conformational changes during natural catalytic activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9271
5-(Iodoacetamido)fluorescein, ≥90% (HPLC)
C22H14INO6