EMAIL THIS PAGE TO A FRIEND
Drug discovery today
A 'rule of three' for fragment-based lead discovery?
PMID 14554012
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
DYNA001
DyNAbind® DNA-Encoded Fragment Library