EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
Antioxidant properties of captopril in vitro.
PMID 14556487
Abstract
This paper reports on the effect of captopril on the stability of unsaturated fatty acids in the evening primrose oil. The experiment was performed for captopril at concentrations of 0.05%, 0.1% and 0.2% in the samples of evening primrose oil at temperatures of 20 degrees, 40 degrees and 60 degrees C. The determination was performed on the 10th day of incubation. The results were compared to those obtained for analogous samples with octyl gallate, an antioxidant commonly used in oil systems, taken at the same concentrations and at the same temperatures. The changes in concentrations of the substances studied with time were monitored by gas chromatography. The results have shown captopril to be a stronger antioxidant than octyl gallate at all concentrations and temperatures used.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48700
Octyl gallate, antioxidant, ≥99.0% (HPLC)
C15H22O5
06717
Octyl gallate, analytical standard
C15H22O5