EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Regioselective bond cleavage in the dissociative electron transfer to benzyl thiocyanates.
PMID 14558793
Abstract
The electrochemical reduction of benzyl thiocyanate and p-nitrobenzyl thiocyanate was investigated in acetonitrile at an inert electrode. These two compounds reveal a change in the reductive cleavage mechanism, and more interestingly, they show a clear-cut example of a regioselective bond dissociation. Both phenomena may be understood on the basis of the dissociative ET theory and its extension to the formation/dissociation reactions of radical ions. While the effect of the standard oxidation potential of the leaving group seems to be predominant in understanding the change in the ET mechanism by changing the driving force, the regioselective cleavage is dictated by changes in the intrinsic barrier related to the nature of the substituent on the aryl moiety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
13929
Benzyl thiocyanate, ≥95.0% (GC)
C8H7NS