EMAIL THIS PAGE TO A FRIEND
Protein and peptide letters
The role of tyrosine residues in the RNA N-glycosidase activity of cinnamomin A-chain.
PMID 14561140
Abstract
Cinnamomin is a type II ribosome-inactivating protein (RIP) and its A-chain (CTA) is a RNA N-glycosidase. It is observed that modification of tyrosine residues by N-acetylimidazole (N-AI) causes almost complete loss of CTA activity. Adenine partially protects tyrosine residues from modification by N-AI. It is proposed that tyrosine residues are involved in the active site of CTA and they are crucial in recognition and binding of ribosomal RNA. Tryptophan residues of CTA are also studied by NBS modification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
157864
1-Acetylimidazole, 98%
C5H6N2O