EMAIL THIS PAGE TO A FRIEND
Biomedicine / [publiee pour l'A.A.I.C.I.G.]
Adrenocortical steroidogenesis and aminoglutethimide I. Biomedical studies.
PMID 14565220
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0496000
Aminoglutethimide, European Pharmacopoeia (EP) Reference Standard
C13H16N2O2