EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
A new class of Spiegelmers containing 2'-fluoro-nucleotides.
PMID 14565337
Abstract
Synthesis of 2'-fluoro-nucleosides from L-arabinose in order to perform the synthesis of 2'-fluoro-Spiegelmers binding to a neuropeptide.