EMAIL THIS PAGE TO A FRIEND
Gene therapy
Creation of immune 'stealth' genes for gene therapy through fusion with the Gly-Ala repeat of EBNA-1.
PMID 14566361
Abstract
A major obstacle in gene-therapy protocols is T-cell-mediated destruction of transgene-expressing cells. Therefore new approaches are needed to prevent rapid clearance of transduced cells. We exploited the Gly-Ala repeat (GAr) domain of the Epstein-Barr virus nuclear antigen-1, since the GAr prevents cytotoxic T-lymphocyte-epitope generation. Here we show that three different enzymes (viz. the E. coli LacZ gene encoded beta-galactosidase, firefly luciferase, and HSV1 thymidine kinase) fused with the GAr retained their function. Moreover, linking GAr with beta-galactosidase successfully prevented recognition of GAr-LacZ-expressing cells by beta-galactosidase-specific CTL. Nonetheless, vaccination with a GAr-LacZ adenovirus or with an allogeneic cell line expressing GAr-LacZ resulted in the induction of beta-gal-specific CTL. This demonstrates that the GAr domain does not inhibit cross presentation of antigens, but only affects breakdown of endogenously synthesized proteins. These data demonstrate how the GAr domain can be exploited to create immuno'stealth' genes by hiding transgene products from CTL-mediated immune attack.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50150
Gly-Ala, ≥99.0% (NT)
C5H10N2O3