EMAIL THIS PAGE TO A FRIEND
European journal of nutrition
Fatty acid profiles, antioxidant status, and growth of preterm infants fed diets without or with long-chain polyunsaturated fatty acids. A randomized clinical trial.
PMID 14569405
Abstract
Long-chain polyunsaturated fatty acids (LCP) are considered conditionally essential nutrients for the infant born prematurely, and attempts are being made to match fatty acid profiles of formula and breast fed infants. In this double-blind, randomized study we investigated the effects of a formula enriched with both n-6 and n-3 LCP on plasma fatty acid profiles, antioxidant status and growth of premature infants. 29 infants received either a formula devoid of LCP or a LCP supplemented formula (0.5 g/100 g fat linoleic acid metabolites, 0.8 g/100 g fat alpha-linolenic acid metabolites). 17 breast fed infants served as a control group. At study entry as well as two and four weeks later, plasma and urine samples were collected, growth data obtained and food tolerance was documented. At the end of the four week study period, plasma docosahexaenoic acid (DHA) levels of supplemented infants were significantly higher than those of unsupplemented infants and similar to those of infants fed human milk. Plasma n-6 LCP concentrations including arachidonic acid (AA) were similar between groups. The plasma alpha-tocopherol levels of breast fed and supplemented infants were similar and tended to be lower than in infants fed the formula devoid of LCP. Urinary malondialdehyde (MDA) excretion of formula fed infants was significantly higher compared to infants fed human milk, but did not differ between the two formula groups. Parameters of growth and milk tolerance did not differ between groups. Our results demonstrate that plasma LCP levels similar to those of breast fed infants can be achieved with the LCP supplemented formula used in this trial, without evidence of adverse effects of the LCP enrichment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2378
γ-Linolenic acid, ≥99%, liquid
C18H30O2
62174
γ-Linolenic acid, analytical standard
C18H30O2