EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthetic studies toward FR182877. Remarkable solvent effect in the vinylogous morita-baylis-hillman cyclization.
PMID 14572290
Abstract
[reaction: see text]. The intramolecular vinylogous Morita-Baylis-Hillman reaction was explored to access the central cyclopentane ring of FR182877. The reaction manifold and product distribution is strikingly solvent and substrate dependent.