EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Double diastereoselection in aldol reactions mediated by dicyclohexylchloroborane between L-erythrulose derivatives and chiral aldehydes. The Felkin-Anh versus Cornforth dichotomy.
PMID 14575488
Abstract
Both matched and mismatched diastereoselections have been observed in aldol reactions of the B,B-dicyclohexylboron enolate of a protected l-erythrulose derivative with a range of chiral aldehydes. The stereochemical outcome of reactions with alpha-methyl aldehydes can be adequately explained within the Felkin-Anh paradigm. In the case of alpha-oxygenated aldehydes, however, strict adherence to this model does not allow for a satisfactory account of the observed results. In such cases, the Cornforth model provides a much better explanation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56845
L-(+)-Erythrulose, ≥85% (HPLC)
C4H8O4