EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
The first occurrence in nature of two compounds from hops.
PMID 14577623
Abstract
Two compounds, (p-methoxyphenyl) diphenylmethanol (1) and tribenzylamine (2), were isolated from Humulus lupulus. Their structures were established on the basis of spectral evidence (MS, IR, NMR, HMBC, HMQC, 1H-1H COSY experiments). Compounds 1 and 2 were found as natural products at the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90660
Tribenzylamine, ≥99.0% (NT)
C21H21N