EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Probing the binding specificity of C-type lectins in vivo.
PMID 14579569
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3879
L-Tyrosinamide
C9H12N2O2