EMAIL THIS PAGE TO A FRIEND
The Tohoku journal of experimental medicine
Involvement of poly(ADP-ribose) synthetase in acoustic trauma of the cochlea.
PMID 14580150
Abstract
We investigated effects of poly(ADP-ribose) synthetase (PARS) inhibitors on acoustic trauma. Albino guinea pigs were intravenously given 3-aminobenzamide, nicotinamide or 3-aminobenzoic acid (an inactive analog of 3-aminobenzamide) just prior to exposure to a 2 kHz pure tone of 120 dB sound pressure level (SPL) for 10 minutes. The threshold of the compound action potential (CAP) and the amplitude of distortion-product otoacoustic emissions (DPOAEs) were measured before and 4 hours after the acoustic overexposure. Statistically significant decreases in the CAP threshold shifts and significant increases in the DPOAE amplitudes were observed 4 hours after the acoustic overexposure in the animals treated with 3-aminobenzamide or nicotinamide, whereas 3-aminobenzoic acid did not exert any protective effect. These results strongly suggest that excessive activation of PARS is involved in generation of the acoustic trauma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
127671
3-Aminobenzoic acid, 98%
C7H7NO2