EMAIL THIS PAGE TO A FRIEND
The Cochrane database of systematic reviews
Erythromycin for the prevention of chronic lung disease in intubated preterm infants at risk for, or colonized or infected with Ureaplasma urealyticum.
PMID 14583992
Abstract
Controversy exists over whether or not Ureaplasma urealyticum colonization or infection of the respiratory tract contributes to the severity of chronic lung disease (CLD), a major cause of morbidity and mortality in preterm infants. To evaluate the efficacy and safety of prophylactic or therapeutic erythromycin in preventing chronic lung disease in intubated preterm infants with unknown U. urealyticum status or proven positivity. Searches were done of MEDLINE (1966-June 9, 2003), EMBASE (1980-May 5, 2003), The Cochrane Central Register of Controlled Trials (CENTRAL, The Cochrane Library, Issue 4, 2002), previous reviews including cross-references, and abstracts of conference proceedings (Pediatric Academic Societies 2000-2003, American Thoracic Society 2001-2003). There were no language restrictions. Expert informants were contacted. Randomized or quasi-randomized studies comparing either prophylactic or therapeutic administration of oral or intravenous erythromycin (regardless of dose and duration) versus no treatment or placebo among intubated preterm infants <37 weeks and <2500 grams with either unknown U. urealyticum status or proven positivity by culture or polymerase chain reaction. Data were extracted by all of the authors independently and differences were resolved by consensus. Treatment effects for categorical outcomes were expressed as relative risk, with 95% confidence intervals. Two small controlled studies, both involving intubated babies <30 weeks gestation, were eligible for inclusion. Lyon 1998 tested prophylactic erythromycin in babies whose U. urealyticum status was unknown at the time of initiation of treatment. Jonsson 1998 tested erythromycin in babies known to be culture positive for U. urealyticum. Neither trial showed a statistically significant effect of erythromycin on CLD, death or the combined outcome CLD or death. Because the two studies differed importantly in their design, the results were not combined in meta-analyses. No adverse effects of a 7-10 day course of erythromycin were reported in either study. Current evidence does not demonstrate a reduction in CLD/death when intubated preterm infants are treated with erythromycin prophylactically before U. urealyticum culture/PCR results are known or when Ureaplasma colonized, intubated preterm infants are treated with erythromycin. However, a true benefit could easily have been missed with the small sample sizes in the two eligible studies. The studies were greatly underpowered to detect uncommon adverse effects such as pyloric stenosis. Additional controlled trials are required to determine whether antibiotic therapy of Ureaplasma reduces CLD and/or death in intubated preterm infants.