EMAIL THIS PAGE TO A FRIEND
Journal of chemical ecology
Factors accounting for the variability in the behavioral response of the onion fly (Delia antiqua) to n-dipropyl disulfide.
PMID 14584680
Abstract
The onion fly Delia antiqua is a specialist herbivore attacking only onions and closely related Allium species. n-Dipropyl disulfide (Pr2S2) has long been known to be attractive to onion flies in the laboratory and in the field. However, the insect's response is highly variable. Using behavioral bioassays we found that Pr2S2 was highly attractive to gravid, mated female onion flies,but did not stimulate oviposition. The response of female onion flies was concentration dependent. The physiological state of the flies (i.e., age, mating status, egg load/oviposition experience) also affected their responsiveness. The response of both sexes of onion flies varied with age, but females were always more strongly attracted than males. Responsiveness of females increased during the first 10 days after emergence. It stayed at a high level until 21 days after which the experiment was terminated. The responsiveness of males reached a maximum at 6-7 days after which it declined. Mated, gravid females responded more strongly to Pr2S2 than unmated, gravid females. Females deprived of the opportunity to oviposit were more attracted compared to females that had oviposited on cut onions prior to the experiment. Electroantennograms (EAG) of females revealed a higher response to stimulation compared to males. The EAG-response of females was not affected by mating status.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
149225
Dipropyl disulfide, 98%
C6H14S2
W322806
Propyl disulfide, ≥97%, FG
C6H14S2