EMAIL THIS PAGE TO A FRIEND
Cancer detection and prevention
Multifactorial activities of nonsteroidal antiestrogens against leukemia.
PMID 14585326
Abstract
The antileukemic activity of nonsteroidal antiestrogens was investigated. Tamoxifen, clomiphene and nafoxidine caused a decrease in viability of the estrogen receptor-negative T-lymphoblastic leukemia cell line CCRF/CEM, nafoxidine being the most active. A combination of clomiphene and genistein resulted in a synergistic cytotoxic effect when applied to Molt-3, another T-lymphblastic leukemic cell line. The antiestrogens arrested the cells at G(0)/G(1) phase and induced apoptosis. Using the CCRF/VCR(1000) cell line, which is resistant to vincristine, it was observed that the effect of nafoxidine on modulating drug resistance was manifested at a lower concentration than that causing a direct cytotoxic effect. Nafoxidine inhibited the Pgp pump activity as measured by rhodamine 123 efflux. Combination with verapamil was found to be more effective in abrogating the pump activity. This study points to the multifactorial activities of nonsteroidal antiestrogens against lymphoblastic leukemia and implies their potential use in clinical treatment as antileukemic drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0118
Nafoxidine hydrochloride, ≥98% (HPLC)
C29H31NO2 · HCl