EMAIL THIS PAGE TO A FRIEND
Toxicologic pathology
Correspondence: Chromotrope 2R.
PMID 14585745
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
202088
Chromotrope 2R, Dye content 75 %
C16H10N2Na2O8S2
C3143
Chromotrope 2R, suitable for modified Gomori Trichrome stain
C16H10N2Na2O8S2