EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
Toward fluorescence nanoscopy.
PMID 14595362
Abstract
For more than a century, the resolution of focusing light microscopy has been limited by diffraction to 180 nm in the focal plane and to 500 nm along the optic axis. Recently, microscopes have been reported that provide three- to sevenfold improved axial resolution in live cells. Moreover, a family of concepts has emerged that overcomes the diffraction barrier altogether. Its first exponent, stimulated emission depletion microscopy, has so far displayed a resolution down to 28 nm. Relying on saturated optical transitions, these concepts are limited only by the attainable saturation level. As strong saturation should be feasible at low light intensities, nanoscale imaging with focused light may be closer than ever.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92546 Abberior® CAGE 500, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C32H22F6N6O5
44254 Abberior® CAGE 500, NHS ester, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C30H19F6N5O6
95705 Abberior® CAGE 532, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C38H34F6N6O7
92545 Abberior® CAGE 552, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C32H28N6O5
94822 Abberior® CAGE 552, NHS ester, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C30H25N5O6
92544 Abberior® FLIP 565, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
79189 Abberior® FLIP 565, NHS ester, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
38361 Abberior® STAR 440SXP, maleimide, for long Stokes STED and 2-color STED application
68221 Abberior® STAR 440SXP, NHS ester, for long Stokes STED and 2-color STED application
94716 Abberior® STAR 470SXP, NHS ester, for STED application
61048 Abberior® STAR 488, NHS ester, for STED application
03004 Abberior® STAR 512, maleimide, for STED application
38922 Abberior® STAR 512, NHS ester, for STED application
38377 Abberior® STAR 580, NHS ester, for STED application
96013 Abberior® STAR 635, maleimide, for STED application
30558 Abberior® STAR 635, NHS ester, for STED application
53366 Anti-Mouse IgG−Abberior® STAR 488 antibody produced in goat, for STED application
52944 Anti-Rabbit IgG−Abberior® STAR 488 antibody produced in goat, for STED application