EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
FRET imaging.
PMID 14595367
Abstract
Förster (or Fluorescence) Resonance Energy Transfer (FRET) is unique in generating fluorescence signals sensitive to molecular conformation, association, and separation in the 1-10 nm range. We introduce a revised photophysical framework for the phenomenon and provide a systematic catalog of FRET techniques adapted to imaging systems, including new approaches proposed as suitable prospects for implementation. Applications extending from a single molecule to live cells will benefit from multidimensional microscopy techniques, particularly those adapted for optical sectioning and incorporating new algorithms for resolving the component contributions to images of complex molecular systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72464
Atto 565 NHS ester, BioReagent, suitable for fluorescence, ≥70.0% (coupling to amines)
C35H34ClN3O11