EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
NMR constrained solution structures for laminin peptide 11. Analogs define structural requirements for inhibition of tumor cell invasion of basement membrane matrix.
PMID 1460013
Abstract
Peptide 11, CDPGYIGSR-NH2, is a segment of laminin which blocks tumor cell invasion. A high affinity laminin receptor in tumor cells is thought to be blocked by the carboxyl-terminal YIGSR, and conformational energy calculations suggest that the glycine in YIGSR allows an important conformational bend. We replaced the YIGSR glycine residue in peptide 11 with either D-alanine or L-alanine to allow or disfavor the proposed glycine bend. We found the Gly7-->D-Ala7 analog to be equal to peptide 11 in inhibiting tumor cell invasion of basement membrane matrix. The Gly7-->L-Ala7 analog was much less capable of invasion inhibition. Two-dimensional 1H-1H NMR was used to study the solution conformations of the peptide 11 analogs. NOESY experiments revealed close NH-NH contacts in peptide 11 and the D-Ala7 analog, but not in the L-Ala7 analog. Molecular dynamics generated low energy structures with excellent NOE agreement for peptide 11 and its analogs. Both peptide 11 and the D-Ala7 analog, but not the less active L-Ala7 analog, were predicted to have similar bends around Gly7 or D-Ala7. These results suggest that a bend in the YIGSR region of peptide 11 may be important for the binding of laminin to its metastasis-associated receptor.