EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Bifunctional agents for reperfusion arrhythmias: novel hybrid vitamin E/class I antiarrhythmics.
PMID 14604685
Abstract
We have synthesized a series of hybrid compounds combining the pharmacophoric redox moieties of vitamin E and key features responsible for the antiarrhythmic properties of the class I antiarrhythmics procainamide and lidocaine. Procainamide analogue (2a) and lidocaine analogues (14a) and (14b) are very strong inhibitors of lipid peroxidation. All analogues tested at 100 or 30 microM enhanced the post ischemic recovery without inducing ventricular fibrillations while there was no evidence in our experiments for drug-induced pro-arrhythmia. In addition, they induced a widening of the QRS intervals. Our data suggest that the efficacy of the new compounds in preventing reperfusion arrhythmias could be attributed to their combined effects involving inhibition of free radical mediated damage coupled with antiarrhythmic properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
359297
Sodium ethanethiolate, technical grade, 80%
C2H5NaS